missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Yeast 100

Product Code. 16249841
Click to view available options
Quantity:
100 g
Unit Size:
100g
This item is not returnable. View return policy

Product Code. 16249841

Brand: Gibco™ 215384

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Yeast 100 is designed to enhance the content of vitamins and other key nutritional components found in yeast.

Yeast 100, is also part of the Peptone Preview Pak which contains:
• Wheat 100, UF—an ultra-filtered supplement and good source of glutamine, which makes it ideal for mammalian cell culture applications. It is digested for enhanced peptide content resulting in a low concentration of free amino acids and carbohydrates.
• Cotton 100, UF—rich in nucleoside content and digested to enhance peptide content. It is processed to provide higher concentrations of vitamins, which can help maintain performance in mammalian and microbial processes.
• Cotton 200, UF—ultra-filtered and rich in nucleoside content, processed to enhance carbohydrate concentrations, which is important for some cell culture and microbial processes
• Pea 100, UF—an ultra-filtered peptone digested to enhance nucleoside and free amino acid content. The high total amino acid content and low carbohydrate content are similar to many high-performance animal origin peptones.
• Soy 100—a highly digested soy protein peptone that is high in nucleosides and free amino acid content. It has high concentrations of vitamins and carbohydrates and is recommend for a wide variety of processes.
• Yeast 100—designed to enhance the content of vitamins and other key nutritional components found in yeast. It is intended for nutritional supplementation for mammalian, microbial, and insect cell culture.

Peptone Preview Pak supplements are non-GMP prototype products that are not for manufacture use. Larger sizes are available through the Gibco Custom Products Services.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Storage conditions: 2°C to 25°C. Store in cool, dry conditions in well-sealed containers.
Quantity 100 g
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.