missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ YDJC Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA566359
This item is not returnable.
View return policy
Description
Immunogen sequence: RVLEGTLAGHT LTAELMAHPG YPSVPPTGGC GEGPDAFSCS WERLHELRVL TAPTLRAQLA QDGVQLCALD DLDSKRPGEE VPCEPTLEPF Highest antigen sequence identity to the following orthologs - mouse 79%, rat 79%.
Probably catalyzes the deacetylation of acetylated carbohydrates an important step in the degradation of oligosaccharides.
Specifications
| YDJC | |
| Polyclonal | |
| Unconjugated | |
| YDJC | |
| 1810015A11Rik; 4930521M19Rik; Carbohydrate deacetylase; UPF0249 protein; UPF0249 protein ydjC homolog; YDJC; YdjC homolog (bacterial) | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 150223 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| A8MPS7 | |
| YDJC | |
| Recombinant Human YDJC. Recombinant protein control fragment (Product #RP-107038). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction