missing translation for 'onlineSavingsMsg'
Learn More

YC/T 184-2004 Labelled Nitrosamines Mixture 690 100 μg/mL in Dichloromethane, Dr. Ehrenstorfer

Product Code. 30308868
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30308868 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30308868 Supplier Dr. Ehrenstorfer Supplier No. DREA50000690DI
 Restricted Item
Estimated Shipment: 07-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (R,S)-N-Nitroso Anatabine-2,4,5,6-d4∣(RS)-N-Nitrosoanabasine pyridinyl-d4∣N-Nitrosonornicotine 2,4,5,6 D4 (pyridine D4)∣N-Nitrosonornicotine-ketoneD4(pyridylD4)
CAS 1020719-69-0, 1020719-68-9, 66148-19-4, 764661-24-7
Solvent or Matrix Dichloromethane
Quantity 1 mL
Concentration or Composition (by Analyte or Components) (R,S)-N-Nitroso Anatabine-2,4,5,6-d4 100 μg/mL∣(RS)-N-Nitrosoanabasine pyridinyl-d4 100 μg/mL∣N-Nitrosonornicotine 2,4,5,6 D4 (pyridine D4) 100 μg/mL∣N-Nitrosonornicotine-ketoneD4(pyridylD4) 100 μg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.