missing translation for 'onlineSavingsMsg'
Learn More
Learn More
YBX2, Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit Polyclonal Antibody
Brand: Abnova H00051087-D01P.100ug
This item is not returnable.
View return policy
Description
Sequence: MSEVEAAAVATAVPAATVPATAAGVVAVVVPVPAGEPQKGGGAGGGGGAASGPAAGTPSAPGSRTPGNPATAVSGTPAPPARSQADKPVLAIQVLGTVKWFNVRNGYGFINRNDTKEDVFVHQTAIKRNNPRKFLRSVGDGETVEFDVVEGEKGAEATNVTGPGGVPVKGSRYAPNRRKSRRFIPRPPSVAPPPMVAEIPSAGTGPGSKGERAEDSGQRPRRWCPPPFFYRRRFVRGPRPPNQQQPIEGTDRVEPKETAPLEGHQQQGDERVPPPRFRPRYRRPFRPRPRQQPTTEGGDGETKPSQGPADGSRPEPQRPRNRPYFQRRRQQAPGPQQAPGPRQPAAPETSAPVNSGDPTTTILESpecifications
| YBX2 | |
| Polyclonal | |
| In 1x PBS, pH 7.4 | |
| NM_015982.1 | |
| YBX2 | |
| YBX2 (NP_057066.1, 1 a.a. ∼ 364 a.a) full-length human protein. | |
| RUO | |
| 51087 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Purified |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Y box binding protein 2 | |
| CONTRIN/CSDA3/DBPC/MGC45104/MSY2 | |
| Rabbit | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction