missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ YBX2 Polyclonal Antibody
GREENER_CHOICE

Product Code. 12829162
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12829162 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12829162 Supplier Invitrogen™ Supplier No. PA521014

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

A suggested positive control is human testis tissue lysate. PA5-21014 can be used with blocking peptide PEP-1128.

YBX2 is a germ-cell specific member of the vertebrate Y box family of nucleic acid-binding proteins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen YBX2
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene YBX2
Gene Accession No. Q9Y2T7, Q9Z2C8
Gene Alias Contrin; CSDA3; Dbpc; DNA-binding protein C; FRGY2 homolog; germ cell specific Y-box binding protein; germ cell-specific Y-box-binding protein; MSY2; MSY2 homolog; RGD1305068; Y box binding protein 2; Y box protein 2; Y-box binding protein 2; Y-box protein MSY2; Y-box-binding protein 2; Ybx2
Gene Symbols YBX2
Host Species Rabbit
Immunogen An 18 amino acid peptide near the carboxy end of human YBX2.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 303250, 51087, 53422
Target Species Human, Mouse, Rat
Content And Storage Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.