missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ YBX1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA519453
This item is not returnable.
View return policy
Description
For Western Blot, this antibody has non-specific bands at 50 kDa. This antibody is predicted to react with Xenopus laevis based on sequence homology. Store antibody at 4ºC for 1-2 weeks. For long-term storage, store at -20ºC.
YBX1 binds to splice sites in pre-mRNA and regulates splice site selection. This protein binds and stabilizes cytoplasmic mRNA and contributes to the regulation of translation by modulating the interaction between the mRNA and eukaryotic initiation factors. It binds to promoters that contain a Y-box (5'-CTGATTGGCCAA-3'), such as HLA class II genes. It regulates the transcription of numerous genes and promotes separation of DNA strands that contain mismatches or are modified by cisplatin. It has endonucleolytic activity and can introduce nicks or breaks into double-stranded DNA (in vitro), and it may play a role in DNA repair.
Specifications
| YBX1 | |
| Polyclonal | |
| Unconjugated | |
| YBX1 | |
| 1700102N10Rik; BP8; BP-8; Byb1; C79409; Cbfa; CBF-A; CCAAT-binding; CCAAT-binding transcription factor 1 subunit A; CCAAT-binding transcription factor I subunit A; CSDA2; CSDB; dbpB; DNA binding protein B; DNA-binding protein B; EF1A; EFI-A; enhancer factor 1 alpha; enhancer factor 1 subunit A; enhancer factor I subunit A; major histocompatibility complex, class II, Y box-binding protein I; MDR NF1; MDR-NF1; MGC104858; MGC110976; MGC117250; Msy1; Msy-1; mYB-1a; Nsep1; NSEP-1; nuclease sensitive element binding protein 1; nuclease-sensitive element-binding protein 1; Y box binding protein 1; Y box protein 1; Y box transcription factor; YB1; YB-1; Y-box binding protein 1; Y-box transcription factor; Y-box-binding protein 1; YBX1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 4904 | |
| -20°C or -80°C if preferred | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1.0 mg/mL | |
| PBS with 0.02% sodium azide; pH 7.4 | |
| P67809 | |
| YBX1 | |
| Synthetic peptide conjugated to KLH derived from within residues 1 - 100 of Human YB1. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction