missing translation for 'onlineSavingsMsg'
Learn More
Learn More
YBX1, Mouse anti-Human, Clone: 4F12, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00004904-M01.100ug
This item is not returnable.
View return policy
Description
Sequence: DKKVIATKVLGTVKWFNVRNGYGFINRNDTKEDVFVHQTAIKKNNPRKYLRSVGDGETVEFDVVEGEKGAEAANVTGPGGVPVQGSKYASpecifications
| YBX1 | |
| Monoclonal | |
| Unconjugated | |
| Y box binding protein 1 | |
| BP-8/CSDA2/CSDB/DBPB/MDR-NF1/MGC104858/MGC110976/MGC117250/NSEP-1/NSEP1/YB-1/YB1 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Immunofluorescence, Western Blot | |
| 4F12 | |
| In 1x PBS, pH 7.4 | |
| NM_004559 | |
| YBX1 | |
| YBX1 (NP_004550, 51 a.a. ∼ 139 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 4904 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction