missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ YARS2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA558513
This item is not returnable.
View return policy
Description
Immunogen sequence: TSTTGAKLGK SAGNAVWLNR DKTSPFELYQ FFVRQPDDSV ERYLKLFTFL PLPEIDHIMQ LHVKEPERRG PQKRLAAEVT KLVHGREG Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 92%.
This gene encodes a mitochondrial protein that catalyzes the attachment of tyrosine to tRNA(Tyr). Mutations in this gene are associated with myopathy with lactic acidosis and sideroblastic anemia type 2 (MLASA2).
Specifications
| YARS2 | |
| Polyclonal | |
| Unconjugated | |
| YARS2 | |
| 2210023C10Rik; CGI-04; EC 6.1.1.1; fi41d10; LOC556702 protein; LOW QUALITY PROTEIN: tyrosine--tRNA ligase, mitochondrial; MLASA2; MT-TYRRS; RGD1311696; tyrosine tRNA ligase 2, mitochondrial; tyrosine--tRNA ligase, mitochondrial; tyrosyl trna synthetase; Tyrosyl-tRNA synthetase; tyrosyl-tRNA synthetase 2; tyrosyl-tRNA synthetase 2 (mitochondrial); tyrosyl-tRNA synthetase 2, mitochondrial; tyrosyl-tRNA synthetase mitochondrial-like protein; tyrosyl-tRNA synthetase, mitochondrial; tyrRS; wu:fi41d10; Yars2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 51067 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9Y2Z4 | |
| YARS2 | |
| Recombinant protein corresponding to Human YARS2. Recombinant protein control fragment (Product #RP-97802). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction