missing translation for 'onlineSavingsMsg'
Learn More

YARS Antibody, Novus Biologicals™

Product Code. 18403631 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
25 μL
Förpackningsstorlek:
0.10mL
25µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Quantity unitSize
18403631 25 μL 25µL
18234607 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 18403631 Leverantör Novus Biologicals Leverantörsnummer NBP18688925ul

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

Rabbit Polyclonal Antibody

YARS Polyclonal specifically detects YARS in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockdown Validated.
TRUSTED_SUSTAINABILITY

Specifikationer

Antigen YARS
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 0.04 - 0.4 ug/mL, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence 0.25-2 ug/mL, Immunohistochemistry-Paraffin 1:50 - 1:200, Knockdown Validated
Formulation PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
Gene Alias CMTDIC, EC 6.1.1, EC 6.1.1.1, Tyrosyl--tRNA ligase, tyrosyl-tRNA synthetase, tyrosyl-tRNA synthetase, cytoplasmic, TyrRS, YRStyrRS, YTStyrosine tRNA ligase 1, cytoplasmic
Gene Symbols YARS1
Host Species Rabbit
Immunogen This antibody was developed against Recombinant Protein corresponding to amino acids:GNVENNGVLSFIKHVLFPLKSEFVILRDEKWGGNKTYTAYVDLEKDFAAEVVHPGDLKNSVEVALNKLLDPIREKFNTPALKKLASAAYPDPSKQKPMAKGPAKNSEPEEVIPSRLDIRVGKIITVEKHPDAD
Purification Method Affinity Purified
Quantity 25 μL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 8565
Test Specificity Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human, Mouse, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Visa mer Visa mindre

For Research Use Only

Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.