missing translation for 'onlineSavingsMsg'
Learn More
Learn More
YAP1 Rabbit anti-Human, Mouse, Zebrafish, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | YAP1 |
|---|---|
| Concentration | 0.3 mg/mL |
| Applications | Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Flow Cytometry, Immunocytochemistry |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16821874
|
Proteintech
13584-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16811874
|
Proteintech
13584-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This antibody is a rabbit polyclonal antibody. The calculated molecular weight of YAP1 is 54 kDa, but phosphorylated YAP1 is about 65 kDa.
YAP1 (Yes-Associated Protein) is a founding member of the WW domain family of proteins, transcriptional co-activator and oncogene. Its gene is transcriptionally activated in squamous cell carcinoma cells. YAP can act both as a coactivator and corepressor and is the critical downstream regulatory target in the Hippo signaling pathway. The Hippo signaling pathway plays a pivotal role in organ size control and tumor suppression by restricting proliferation and promoting apoptosis. Also plays key roles in tissue tension, 3D tissue shape, cell proliferation, cell death, and cell migration. Mutations in the IRF5 gene results in coloboma.Specifications
| YAP1 | |
| Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Flow Cytometry, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Zebrafish, Human, Mouse | |
| P46937, P46938, Q1L8J7 | |
| 10413, 22601, 561411 | |
| YAP1 Fusion Protein Ag4510 | |
| Primary | |
| -20°C | |
| YAP1 |
| 0.3 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| 65 kDa Yes associated protein, YAP, YAP1, YAP2, YAP65, YKI, Yorkie homolog | |
| yap1 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit