missing translation for 'onlineSavingsMsg'
Learn More

YAP1, p Ser61 Antibody, Novus Biologicals™

Product Code. 18368045 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18368045 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18368045 Supplier Novus Biologicals Supplier No. NBP313319

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

YAP1 Polyclonal antibody specifically detects YAP1 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen YAP1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500-1:3000
Formulation PBS, 1% BSA, 20% Glycerol (pH7)
Gene Alias 65 kDa Yes-associated protein, COB1, Protein Yorkie Homolog, Transcriptional Coactivator YAP1, YAP, YAP1-2gamma, YAP2, YAP2L, YAP65, YAP65yes-associated protein 2, Yes Associated Protein, Yes-Associated Protein, Yes-associated protein 1, Yes-associated protein 1, 65kDa, yes-associated protein beta, yes-associated protein delta, Yes-Associated Protein YAP65 Homolog, YKI, yorkie homolog
Host Species Rabbit
Immunogen Carrier-protein conjugated synthetic peptide corresponding to residues around human YAP1 (phospho Ser61). The exact sequence is proprietary.
Purification Method Antigen Affinity-purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Breast Cancer, Cancer, Chromatin Research, Core ESC Like Genes, Signal Transduction, Stem Cell Markers, Transcription Factors and Regulators, Tyrosine Kinases
Primary or Secondary Primary
Gene ID (Entrez) 10413
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.