missing translation for 'onlineSavingsMsg'
Learn More
Learn More
YAP1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£248.00 - £478.00
Specifications
| Antigen | YAP1 |
|---|---|
| Concentration | 0.2 mg/mL |
| Dilution | Western Blot 1:1000, Simple Western 1:12.5, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence, Immunoprecipitation 2-10 ug, Immunohistochemistry-Paraffin 1:50-1:200, Immunohistochemistry-Frozen reported in scientific literature (PMID 28581498), Immunoblotting reported in scientific literature (PMID 28406163), Chromatin Immunoprecipitation (ChIP), Knockout Validated, KnockDown Validated reported in scientific literature (PMID 28406163) |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunoblot, Immunoassay, KnockDown |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18774953
|
Novus Biologicals
NB110-58358SS |
0.025 mL |
£248.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18785923
|
Novus Biologicals
NB110-58358 |
£478.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
Description
YAP1 Polyclonal specifically detects YAP1 in Human, Mouse, Rat, Canine, Zebrafish samples. It is validated for Western Blot, ChIP assay, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunoblot, Immunoassay, Knockdown.Specifications
| YAP1 | |
| Western Blot 1:1000, Simple Western 1:12.5, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence, Immunoprecipitation 2-10 ug, Immunohistochemistry-Paraffin 1:50-1:200, Immunohistochemistry-Frozen reported in scientific literature (PMID 28581498), Immunoblotting reported in scientific literature (PMID 28406163), Chromatin Immunoprecipitation (ChIP), Knockout Validated, KnockDown Validated reported in scientific literature (PMID 28406163) | |
| Polyclonal | |
| Rabbit | |
| Breast Cancer, Cancer, Chromatin Research, Core ESC Like Genes, Signal Transduction, Stem Cell Markers, Transcription Factors and Regulators, Tyrosine Kinases | |
| PBS, 15% glycerol with 0.02% Sodium Azide | |
| 65 kDa Yes-associated protein, COB1, Protein Yorkie Homolog, Transcriptional Coactivator YAP1, YAP, YAP1-2gamma, YAP2, YAP2L, YAP65, YAP65yes-associated protein 2, Yes Associated Protein, Yes-Associated Protein, Yes-associated protein 1, Yes-associated protein 1, 65kDa, yes-associated protein beta, yes-associated protein delta, Yes-Associated Protein YAP65 Homolog, YKI, yorkie homolog | |
| YAP1 | |
| IgG | |
| Affinity Purified | |
| Expected reactivity based on immunogen homology: Isoform 4 (100%), Isoform 6 (100%) |
| 0.2 mg/mL | |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunoblot, Immunoassay, KnockDown | |
| Unconjugated | |
| RUO | |
| Human, Mouse, Rat, Canine, Zebrafish | |
| P46937 | |
| 10413 | |
| This YAP1 Antibody was developed against a partial recombinant human YAP1 protein expressed in bacteria. [Uniprot: P46937], N-terminal GST fusion protein | |
| Primary | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
| 48 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title