missing translation for 'onlineSavingsMsg'
Learn More
Learn More
YAP1 Antibody (1A12), DyLight 650, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-22117C
This item is not returnable.
View return policy
Description
YAP1 Monoclonal specifically detects YAP1 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Knockout Validated, Knockdown Validated.
Specifications
| YAP1 | |
| Monoclonal | |
| DyLight 650 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| YAP1 | |
| Purified recombinant fragment of human YAP1 expressed in E. coli. [UniProt# P46937] | |
| 0.1 mL | |
| Breast Cancer, Cancer, Chromatin Research, Core ESC Like Genes, Signal Transduction, Stem Cell Markers, Transcription Factors and Regulators, Tyrosine Kinases | |
| 10413 | |
| Store at 4C in the dark. | |
| IgG1 |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoassay, KnockDown | |
| 1A12 | |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Knockout Validated, KnockDown Validated | |
| 65 kDa Yes-associated protein, COB1, Protein Yorkie Homolog, Transcriptional Coactivator YAP1, YAP, YAP1-2gamma, YAP2, YAP2L, YAP65, YAP65yes-associated protein 2, Yes Associated Protein, Yes-Associated Protein, Yes-associated protein 1, Yes-associated protein 1, 65kDa, yes-associated protein beta, yes-associated protein delta, Yes-Associated Protein YAP65 Homolog, YKI, yorkie homolog | |
| Mouse | |
| Ammonium sulfate precipitation | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction