missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
YAP1 Monoclonal antibody specifically detects YAP1 in Human samples. It is validated for Western Blot, Simple Western, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Gene Knock-Out, KnockDown
Specifications
Specifications
| Antigen | YAP1 |
| Applications | Western Blot, Simple Western, Flow Cytometry, ELISA, Immunohistochemistry |
| Classification | Monoclonal |
| Clone | 1A12 |
| Conjugate | DyLight 594 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | 65 kDa Yes-associated protein, COB1, Protein Yorkie Homolog, Transcriptional Coactivator YAP1, YAP, YAP1-2gamma, YAP2, YAP2L, YAP65, YAP65yes-associated protein 2, Yes Associated Protein, Yes-Associated Protein, Yes-associated protein 1, Yes-associated protein 1, 65kDa, yes-associated protein beta, yes-associated protein delta, Yes-Associated Protein YAP65 Homolog, YKI, yorkie homolog |
| Host Species | Mouse |
| Immunogen | Purified recombinant fragment of human YAP1 expressed in E. coli. [UniProt# P46937] |
| Purification Method | Ammonium sulfate precipitation |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?