missing translation for 'onlineSavingsMsg'
Learn More

YAF2 Antibody, Biotin, Novus Biologicals™

Product Code. 18433361 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18433361 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18433361 Supplier Novus Biologicals Supplier No. NBP204127B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

YAF2 Polyclonal antibody specifically detects YAF2 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen YAF2
Applications Western Blot
Classification Polyclonal
Conjugate Biotin
Dilution Western Blot
Formulation PBS
Gene Alias DKFZp779H1820, MGC41856, YY1 associated factor 2, YY1-associated factor 2
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 50 and 100 of human YY1 Associated Factor 2 using the numbering given in entry NP_005739.2 (GeneID 10138).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10138
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.