missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Y3 Yellow Color Reference Solution, According to European Pharmacopoeia (EP), Reagecon
Shop All Reagecon ProductsDescription
- Manufactured gravimetrically with certificate of calibration issued in accordance with ISO 17025
- Concentration verified using high performance spectrophotometer
Specifications
Specifications
| Packaging | Amber glass Winchester bottle |
| Certificate of Analysis | Yes |
| Physical Form | Liquid |
| UN Number | 1760 |
| Type | Color Reference Standard |
| Quantity | 100 mL |
Safety and Handling
Signal Word
- Danger
- Carcinogenicity Category 1B
- LONG-TERM AQUATIC HAZARD Chronic 3
- H350i-May cause cancer by inhalation.
- H412-Harmful to aquatic life with long lasting effects.
- P201-Obtain special instructions before use.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contain: hydrochloric acid, cobalt dichloride
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?