missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Y06137, MedChemExpress
Description
Y06137 is a potent and selective BET inhibitor for treatment of castration-resistant prostate cancer (CRPC). Y06137 binds to the BRD4(1) bromodomain with a Kd of 81 nM.
Specifications
Specifications
| Chemical Name or Material | Y06137 |
| CAS | 2226534-49-0 |
| Color | Off-White |
| Purity Grade Notes | Research |
| Molecular Formula | C27H32N4O2 |
| Quantity | 10 mg |
| Solubility Information | DMSO : 62.5 mg/mL (140.59 mM; Need ultrasonic) |
| SMILES | CC1=NOC(C1=C2)=CC=C2C3=NC(C=C4)=C(N3CC5CCCCC5)C=C4N6CCOC[C@@H]6C |
| Molecular Weight (g/mol) | 444.57 |
| Formula Weight | 444.57 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?