missing translation for 'onlineSavingsMsg'
Learn More

Y06036, MedChemExpress

Product Code. 16490787
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16490787 50 mg 50mg
16464478 10 mg 10mg
16463106 25 mg 25mg
16415356 100 mg 100mg
16407207 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16490787 Supplier Medchem Express Supplier No. HY111502
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Y06036 is a potent and selective BET inhibitor, which binds to the BRD4(1) bromodomain with Kd value of 82 nM. Antitumor activity.

Y06036 is a potent and selective BET inhibitor, which binds to the BRD4(1) bromodomain with Kd value of 82 nM. Antitumor activity.

Specifications

Chemical Name or Material Y06036
CAS 1832671-96-1
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C16H15BrN2O5S
Quantity 50 mg
Solubility Information DMSO : 130 mg/mL (304.26 mM; Need ultrasonic)
SMILES BrC1=CC(S(=O)(NC2=C(OC)C=C(ON=C3C)C3=C2)=O)=C(OC)C=C1
Molecular Weight (g/mol) 427.27
Formula Weight 427.27
Percent Purity 98.96%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.