missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Y-79 (human retinoblastoma) Whole cell lysate, Non-denatured; Abnova
Click to view available options
Quantity:
200 μg
Unit Size:
200µg
Description
- Tissue: Retina
- Host: Human
- Lysis buffer: Modified RIPA lysis buffer: 50mM Tris-HCl pH 7.4, 150mM NaCl, 1mM EDTA, 1% Triton X-100, 0.1% SDS, 1% Sodium deoxycholate, 1mM PMSF
- Storage buffer: In modified RIPA lysis buffer
Western Blotting, Immunoprecipitation
Specifications
Specifications
| For Use With (Application) | Immunoprecipitation, Western Blot |
| Tissue | Retina |
| Description | Whole cell lysate (non-denatured) |
| Lysis Buffer | Modified RIPA Lysis Buffer: 50 mM Tris-HCl pH 7.4, 150 mM NaCl, 1mM EDTA, 1% Triton X-100, 0.1% SDS, 1% Sodium deoxycholate, 1mM PMSF. |
| Preparation Method | Cell lysate was prepared by homogenization in ice-cold modified RIPA Lysis Buffer with cocktail of protease inhibitors (Sigma). Cell debris was removed by centrifugation. Protein concentration was determined with Bradford assay (Bio-Rad protein assay, Microplate Standard assay). |
| Quality Control Testing | 12.5% SDS-PAGE Stained with Coomassie Blue. |
| Quantity | 200 μg |
| Storage Buffer | In modified RIPA Lysis Buffer. |
| Host Species | Human |
| Concentration | 5 mg/mL |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction