missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Y-33075, MedChemExpress
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
Y-33075 is a selective ROCK inhibitor derived from Y-27632, and is more potent than Y-27632, with an IC50 of 3.6 nM.
Specifications
Specifications
| Chemical Name or Material | Y-33075 |
| CAS | 199433-58-4 |
| Color | Off-White |
| Purity Grade Notes | Research |
| Molecular Formula | C16H16N4O |
| Quantity | 10 mg |
| Synonym | Y 39983 |
| Solubility Information | DMSO : 50 mg/mL (178.37 mM; Need ultrasonic and warming) ∣H2O : 1 mg/mL (3.57 mM; ultrasonic and heat to 60°C) |
| SMILES | O=C(NC1=C2C(NC=C2)=NC=C1)C3=CC=C(C=C3)[C@H](N)C |
| Molecular Weight (g/mol) | 280.32 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction