missing translation for 'onlineSavingsMsg'
Learn More

Y-320, MedChemExpress

Product Code. 16486908
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16486908 10 mg 10mg
16482967 50 mg 50mg
16419447 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16486908 Supplier Medchem Express Supplier No. HY15898
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Y-320 is a new phenylpyrazoleanilide immunomodulator; inhibits IL-17 production by CD4 T cells stimulated with IL-15 with IC50 values of 20 to 60 nM.

Y-320 is a new phenylpyrazoleanilide immunomodulator

Specifications

Chemical Name or Material Y-320
CAS 288250-47-5
Color Pink
Purity Grade Notes Research
Molecular Formula C27H29ClN6O2
Quantity 10 mg
Solubility Information DMSO : 5.5 mg/mL (10.89 mM; Need ultrasonic)
SMILES O=C(C1=C(C)N(C2=CC=C(Cl)C=C2)N=C1)NC3=CC=C(N4CCC(N5CCOCC5)CC4)C(C#N)=C3
Molecular Weight (g/mol) 505.01
Formula Weight 505.01
Percent Purity 99.39%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.