missing translation for 'onlineSavingsMsg'
Learn More

Y-320, MedChemExpress

Product Code. 16419447
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16419447

Brand: Medchem Express HY158985mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Y-320 is a new phenylpyrazoleanilide immunomodulator; inhibits IL-17 production by CD4 T cells stimulated with IL-15 with IC50 values of 20 to 60 nM.

Y-320 is a new phenylpyrazoleanilide immunomodulator

Specifications

Chemical Name or Material Y-320
CAS 288250-47-5
Color Pink
Purity Grade Notes Research
Molecular Formula C27H29ClN6O2
Quantity 5 mg
Solubility Information DMSO : 5.5 mg/mL (10.89 mM; Need ultrasonic)
SMILES O=C(C1=C(C)N(C2=CC=C(Cl)C=C2)N=C1)NC3=CC=C(N4CCC(N5CCOCC5)CC4)C(C#N)=C3
Molecular Weight (g/mol) 505.01
Formula Weight 505.01
Percent Purity 99.39%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.