missing translation for 'onlineSavingsMsg'
Learn More

Y-27632 dihydrochloride, MedChemExpress

Product Code. 16474058
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
200 mg
500 mg
1 g
2 g
Unit Size:
100mg
10mg
1g
200mg
2g
500mg
50mg
5mg
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16474058 10 mg 10mg
16486757 5 mg 5mg
16485036 100 mg 100mg
16480116 2 g 2g
16460996 200 mg 200mg
16430417 50 mg 50mg
16401586 500 mg 500mg
16401069 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Product Code. 16474058 Supplier Medchem Express Supplier No. HY10583
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Y-27632 dihydrochloride is an orally active, ATP-competitive inhibitor of ROCK-I and ROCK-II, with Kis of 220 and 300 nM, respectively.

Y-27632 dihydrochloride attenuates Doxorubicin-induced apoptosis of human cardiac stem cells. Y-27632 also suppresses dissociation-induced apoptosis of murine prostate stem/progenitor cells. Y-27632 dihydrochloride primes human induced pluripotent stem cells (hIPSCs) to selectively differentiate towards mesendodermal lineage via epithelial-mesenchymal transition-like modulation.

Specifications

Chemical Name or Material Y-27632 dihydrochloride
CAS 129830-38-2
Color White
Purity Grade Notes Research
Molecular Formula C14H23Cl2N3O
Quantity 10 mg
Solubility Information H2O : 100 mg/mL (312.25 mM; Need ultrasonic) ∣DMSO : 33.33 mg/mL (104.07 mM; Need ultrasonic)
SMILES O=C([C@@H]1CC[C@]([C@H](N)C)([H])CC1)NC2=CC=NC=C2.[H]Cl.[H]Cl
Molecular Weight (g/mol) 320.26
Formula Weight 320.26
Percent Purity 95.0%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.