missing translation for 'onlineSavingsMsg'
Learn More

Y-26763, Tocris Bioscience™

Product Code. 18065722
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18065722 50 mg 50mg
18054532 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18065722 Supplier Tocris Bioscience Supplier No. 2076/50

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Kir6 (KATP) channel opener

Y-26763 is a Kir6 (KATP) channel opener and active metabolite of Y-27152. Relaxes contracted rat aortic rings (IC50 = 0.027 mM) and inhibits glucose-induced insulin secretion in isolated human pancreatic β-cells in vitro. Produces hypotension in spontaneously hypertensive rats and displays cardioprotective properties in dogs following systemic administration in vivo.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 127408-31-5
Molecular Formula C14H16N2O4
Molecular Weight (g/mol) 276.292
InChI Key IWTCFIIOUXJOOV-OLZOCXBDSA-N
Synonym unii-q9f3xv8a6x, q9f3xv8a6x, +-3s,4r-n-acetyl-n-hydroxy amino-6-cyano-3,4-dihydro-2,2-dimethyl-2h-1-benzopyran-3-ol, n-3s,4r-6-cyano-3,4-dihydro-3-hydroxy-2,2-dimethyl-2h-1-benzopyran-4-yl-n-hydroxyacetamide, acetamide, n-6-cyano-3,4-dihydro-3-hydroxy-2,2-dimethyl-2h-1-benzopyran-4-yl-n-hydroxy-, 3s-trans, n-3s,4r-6-cyano-3-hydroxy-2,2-dimethylchroman-4-yl-n-hydroxyacetamide, acetamide,n-3s,4r-6-cyano-3,4-dihydro-3-hydroxy-2,2-dimethyl-2h-1-benzopyran-4-yl-n-hydroxy, n-3s,4r-6-cyano-3-hydroxy-2,2-dimethyl-3,4-dihydro-1-benzopyran-4-yl-n-hydroxyacetamide, d05gpx, n-3s,4r-6-cyano-3-hydroxy-2,2-dimethyl-3,4-dihydrochromen-4-yl-n-hydroxyacetamide
PubChem CID 121878
IUPAC Name N-[(3S,4R)-6-cyano-3-hydroxy-2,2-dimethyl-3,4-dihydrochromen-4-yl]-N-hydroxyacetamide
SMILES CC(=O)N(C1C(C(OC2=C1C=C(C=C2)C#N)(C)C)O)O

Specifications

Quantity 50 mg
Formula Weight 276.29
Percent Purity >98%
Chemical Name or Material Y-26763
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.