missing translation for 'onlineSavingsMsg'
Learn More

Y-26763, Tocris Bioscience™

Product Code. 18054532
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18054532 10 mg 10mg
18065722 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18054532 Supplier Tocris Bioscience Supplier No. 2076/10

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Kir6 (KATP) channel opener

Y-26763 is a Kir6 (KATP) channel opener and active metabolite of Y-27152. Relaxes contracted rat aortic rings (IC50 = 0.027 mM) and inhibits glucose-induced insulin secretion in isolated human pancreatic β-cells in vitro. Produces hypotension in spontaneously hypertensive rats and displays cardioprotective properties in dogs following systemic administration in vivo.
TRUSTED_SUSTAINABILITY

Identificadores de productos químicos

CAS 127408-31-5
Molecular Formula C14H16N2O4
Molecular Weight (g/mol) 276.292
InChI Key IWTCFIIOUXJOOV-OLZOCXBDSA-N
Synonym unii-q9f3xv8a6x, q9f3xv8a6x, +-3s,4r-n-acetyl-n-hydroxy amino-6-cyano-3,4-dihydro-2,2-dimethyl-2h-1-benzopyran-3-ol, n-3s,4r-6-cyano-3,4-dihydro-3-hydroxy-2,2-dimethyl-2h-1-benzopyran-4-yl-n-hydroxyacetamide, acetamide, n-6-cyano-3,4-dihydro-3-hydroxy-2,2-dimethyl-2h-1-benzopyran-4-yl-n-hydroxy-, 3s-trans, n-3s,4r-6-cyano-3-hydroxy-2,2-dimethylchroman-4-yl-n-hydroxyacetamide, acetamide,n-3s,4r-6-cyano-3,4-dihydro-3-hydroxy-2,2-dimethyl-2h-1-benzopyran-4-yl-n-hydroxy, n-3s,4r-6-cyano-3-hydroxy-2,2-dimethyl-3,4-dihydro-1-benzopyran-4-yl-n-hydroxyacetamide, d05gpx, n-3s,4r-6-cyano-3-hydroxy-2,2-dimethyl-3,4-dihydrochromen-4-yl-n-hydroxyacetamide
PubChem CID 121878
IUPAC Name N-[(3S,4R)-6-cyano-3-hydroxy-2,2-dimethyl-3,4-dihydrochromen-4-yl]-N-hydroxyacetamide
SMILES CC(=O)N(C1C(C(OC2=C1C=C(C=C2)C#N)(C)C)O)O

Especificaciones

Quantity 10 mg
Formula Weight 276.29
Percent Purity >98%
Chemical Name or Material Y-26763
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.