missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Y-26763, MedChemExpress
Description
Y-26763 is a K+ channel opener and active metabolite of Y-27152. Y-26763 is an ATP-sensitive K+ (KATP) channel activator.
Specifications
Specifications
| Chemical Name or Material | Y-26763 |
| CAS | 127408-31-5 |
| Color | Off-White |
| Purity Grade Notes | Research |
| Molecular Formula | C14H16N2O4 |
| Quantity | 10 mg |
| SMILES | CC(N([C@H]1[C@H](O)C(C)(C)OC2=CC=C(C#N)C=C12)O)=O |
| Molecular Weight (g/mol) | 276.29 |
| Formula Weight | 276.29 |
| Percent Purity | 99.0% |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?