missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Y 134

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
Selective estrogen receptor modulator (SERM) that displays selectivity for ERα over ERβ (Ki values are 0.09 and 11.31 nM respectively) and no cross-reactivity with mineralocorticoid, glucocorticoid, androgen and progesterone receptors. Acts as an agonist in the bone and antagonist in reproductive tissue. Suppresses estrogen-stimulated proliferation of ER-positive human breast cancer MCF-7 and T47D cells.
Specifications
Specifications
| Quantity | 10 mg |
| Formula Weight | 472.6 |
| Purity | >98% |
| Molecular Weight (g/mol) | 472.603 |
| Chemical Name or Material | Y 134 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction