missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ XYLT1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA567627
This item is not returnable.
View return policy
Description
Predicted to react with Mouse and Rat samples.
Catalyzes the first step in biosynthesis of glycosaminoglycan. Transfers D-xylose from UDP-D-xylose to specific serine residues of the core protein. Initial enzyme in the biosynthesis of chondroitin sulfate and dermatan sulfate proteoglycans in fibroblasts and chondrocytes.
Specifications
| XYLT1 | |
| Polyclonal | |
| Unconjugated | |
| Xylt1 | |
| 8030490L12; beta-D-xylosyltransferase 1; DBQD2; Peptide O-xylosyltransferase 1; PXYLT1; Xt1; XTI; XT-I; Xylosyltransferase 1; xylosyltransferase I; xylosyltransferase iota; XYLT1; XYLT-1; XYLTI; xylT-I | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 233781, 64131, 64133 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.7 mg/mL | |
| PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| Q811B1, Q86Y38, Q9EPI1 | |
| Xylt1 | |
| Synthesized peptide derived from human XYLT1. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction