missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Xylometazoline Hydrochloride, Mikromol™
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Description
Xylometazoline Hydrochloride
Specifications
Specifications
| Chemical Name or Material | Xylometazoline Hydrochloride |
| CAS | 1218-35-5 |
| Analyte or Component Names | Xylometazoline Hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C16 H24 N2 . Cl H |
| Certifications/Compliance | ISO 17034 |
| Synonym | Oxymetazoline Hydrochloride Imp. B (EP), Oxymetazoline Imp. B (EP), Xylometazoline Hydrochloride, Oxymetazoline Hydrochloride Imp. B (EP) as Hydrochloride, 2-[[4-(1,1-Dimethylethyl)-2,6-dimethylphenyl]methyl]-4,5-dihydro-1H-imidazole hydrochloride (1:1), 2-[[4-(1,1-Dimethylethyl)-2,6-dimethylphenyl]methyl]-4,5-dihydro-1H-imidazole monohydrochloride, 2-(4-tert-Butyl-2,6-dimethylbenzyl)-2-imidazoline monohydrochloride, Brizolin, Galazolin, Neo-Rinoleina, Neo-Synephrine II, Novorin, Olynth, Otrivin hydrochloride, Otrivine, Otrivine hydrochloride, Therapin, Xymelin, Oxymetazoline Hydrochloride Impurity B as Hydrochloride, Oxymetazoline Impurity B as Hydrochloride |
| InChI Formula | InChI=1S/C16H24N2.ClH/c1-11-8-13(16(3,4)5)9-12(2)14(11)10-15-17-6-7-18-15;/h8-9H,6-7,10H2,1-5H3,(H,17,18);1H |
| SMILES | Cl.Cc1cc(cc(C)c1CC2=NCCN2)C(C)(C)C |
| IUPAC Name | 2-[(4-tert-butyl-2,6-dimethylphenyl)methyl]-4,5-dihydro-1H-imidazole;hydrochloride |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction