missing translation for 'onlineSavingsMsg'
Learn More

Xylitol, LoGiCal

Product Code. 30460780
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Product Code. Quantity unitSize
30460780 10 mg 10mg
1 options
This item is not returnable. View return policy

Product Code. 30460780

Brand: LoGiCal LGCFOR1451.00

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

LoGiCal delivers certified reference materials you can trust as neats, native solutions and stable isotope labelled solutions, with precise documentation and expert support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.

Xylitol
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Xylitol
CAS 87-99-0
Analyte or Component Names Xylitol
Shipping Condition Room Temperature
Molecular Formula C5 H12 O5
Certifications/Compliance ISO 9001
Synonym Xylitol, Meso-xylitol, C-Xylidex CR 16055, DL-Xylitol, Eutrit, Fluorette, Klinit, Kylit, NSC 25283, Wood sugar alcohol, x 0018, Xylisorb, Xylisorb 300, Xylisorb 700, Xylit, Xylit XC, Xylitab 100, Xylitab 300, Xylitab DC, Xylite, Xylite (sugar), Xylitol C, Xylitol CM 90, Xylitol P, Xyliton, meso-Xylitol, xylo-Pentitol
InChI Formula InChI=1 S/C5H12O5/c6-1-3(8)5(10)4(9)2-7/h3-10 H,1-2H2/t3-,4+,5+
SMILES OC[C@@H](O)[C@H](O)[C@@H](O)CO
IUPAC Name (2 S,4 R)-pentane-1,2,3,4,5-pentol
Industry Type Forensics
Molecular Weight (g/mol) 152.15
Quantity 10 mg
Formula Weight 152.0685 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.