missing translation for 'onlineSavingsMsg'
Learn More

Xylitol, 99+%, Thermo Scientific Chemicals

Product Code. 10743455
Click to view available options
Quantity:
25 g
100 g
500 g
2.5 kg
Packaging:
Plastic bottle

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 87-99-0
Molecular Formula C5H12O5
Molecular Weight (g/mol) 152.15
MDL Number MFCD00064291,MFCD00064292
InChI Key HEBKCHPVOIAQTA-UHFFFAOYNA-N
Synonym xylitol, ribitol, adonitol, d-xylitol, xylite, adonit, adonite, xylit, d-ribitol, eutrit
PubChem CID 6912
IUPAC Name (2S,4R)-pentane-1,2,3,4,5-pentol
SMILES OCC(O)C(O)C(O)CO

Specifications

Melting Point 92.0°C to 96.0°C
Color White
Boiling Point 215.0°C to 217.0°C
Infrared Spectrum Authentic
Assay Percent Range 2% max. Other polyols (GC)
Packaging Plastic bottle
Linear Formula HOCH2[CH(OH)]3CH2OH
Quantity 2.5 kg
Beilstein 01, 531
Merck Index 15, 10283
Solubility Information Solubility in water: 1700g/L (20°C). Other solubilities: soluble in pyridine,less soluble in alcohols
Formula Weight 152.15
Percent Purity 99+%
Physical Form Crystalline Powder
Chemical Name or Material Xylitol, 99+%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'