missing translation for 'onlineSavingsMsg'
Learn More

Xylitol, 99+%

Product Code. 10371514 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Unit Size:
100g
2.5kg
25g
500g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10371514 500 g Plastic bottle 500g
10055663 25 g Plastic bottle 25g
10276610 100 g Plastic bottle 100g
10743455 2.5 kg Plastic bottle 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10371514 Supplier Thermo Scientific Acros Supplier No. 225985000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 87-99-0 | C5H12O5 | 152.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 87-99-0
Molecular Formula C5H12O5
Molecular Weight (g/mol) 152.15
MDL Number MFCD00064291,MFCD00064292
InChI Key HEBKCHPVOIAQTA-UHFFFAOYNA-N
Synonym xylitol, ribitol, adonitol, d-xylitol, xylite, adonit, adonite, xylit, d-ribitol, eutrit
PubChem CID 6912
IUPAC Name (2S,4R)-pentane-1,2,3,4,5-pentol
SMILES OCC(O)C(O)C(O)CO

Specifications

Melting Point 92.0°C to 96.0°C
Color White
Boiling Point 215.0°C to 217.0°C
Infrared Spectrum Authentic
Assay Percent Range 2% max. Other polyols (GC)
Packaging Plastic bottle
Linear Formula HOCH2[CH(OH)]3CH2OH
Quantity 500 g
Beilstein 01, 531
Merck Index 15, 10283
Solubility Information Solubility in water: 1700g/L (20°C). Other solubilities: soluble in pyridine,less soluble in alcohols
Formula Weight 152.15
Percent Purity 99+%
Physical Form Crystalline Powder
Chemical Name or Material Xylitol, 99+%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.