missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Kallikrein 11 Monoclonal specifically detects Kallikrein 11 in Human samples. It is validated for Sandwich ELISA Capture.
Specifications
Specifications
| Antigen | Kallikrein 11 |
| Applications | Sandwich ELISA Capture |
| Classification | Monoclonal |
| Clone | 101 |
| Conjugate | APC |
| Dilution | Sandwich ELISA Capture |
| Formulation | PBS |
| Gene Alias | EC 3.4.21, EC 3.4.21.-, EC 3.4.21.4, Hippostasin, hK11, kallikrein 11, kallikrein-related peptidase 11, MGC33060, PRSS20kallikrein-11, Serine protease 20, TLSPhippostasin, Trypsin-like protease |
| Host Species | Rabbit |
| Immunogen | This antibody was obtained from a rabbit immunized with purified, recombinant Human Kallikrein 11 (Accession#: NP_006844.1; Met1-Asn250). |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?