missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Xylenol Orange tetrasodium salt, ACS

Product Code. 11304098
Change view
Click to view available options
Quantity:
2 g
10 g
50 g
Unit Size:
10g
2g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11304098 10 g 10g
11384088 2 g 2g
11394088 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11304098 Supplier Thermo Scientific Chemicals Supplier No. 041379.09
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 3618-43-7

Xylenol Orange tetrasodium salt is used as a photometric reagent and a metal indicator. It is a versatile indicator for EDTA titrations in acidic range. It is a common organic reagent used as an indicator for metal titrations.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3618-43-7
MDL Number MFCD00044293

Specifications

Melting Point 210°C (decomposition)
Solubility Information Soluble in water.
Formula Weight 760.59
Grade ACS
Odor Odorless
Quantity 10 g
Physical Form Powder
Chemical Name or Material Xylenol Orange tetrasodium salt

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.