missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Xylenol orange, sodium salt, ACS reagent

Product Code. 13478288 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
13478288 25 g Glass bottle 25g
13408298 1 g Glass bottle 1g
13418298 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 13478288 Supplier Thermo Scientific Chemicals Supplier No. 447600250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3618-43-7 | C31H28N2Na4O13S | 760.59 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3618-43-7
Molecular Formula C31H28N2Na4O13S
Molecular Weight (g/mol) 760.59

Specifications

Melting Point 210°C
Packaging Glass bottle
Quantity 25 g
Merck Index 15,10281
Solubility Information Solubility in water: 510g/L (20°C)
Formula Weight 760.59
Clarity Passes test
Grade ACS Reagent
Chemical Name or Material Xylenol orange, sodium salt

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.