missing translation for 'onlineSavingsMsg'
Learn More

Xylenes, 99%, ACS reagent

Product Code. 10247273 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
500 mL
2.5 L
25 L
Unit Size:
2.5 L
25 L
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10247273 25 L 25 L
10453931 500 mL 500mL
10741624 2.5 L 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
Product Code. 10247273 Supplier Thermo Scientific Acros Supplier No. 422680250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

CAS: 1330-20-7 | C8H10

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

EcoSafPak™ is an environmentally friendly packaging system made of 100% recyclable material by a SFI certified supplier.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1330-20-7 , 100-41-4
Molecular Formula C8H10
MDL Number MFCD00077264
Synonym Dimethylbenzene

Specifications

Melting Point -34.0°C
Density 0.8650g/mL
Quantity 25 L
Flash Point 23°C
Infrared Spectrum Authentic
Packaging Metal Drum
Physical Form Liquid
Refractive Index 1.4940 to 1.4980
Residue after Evaporation 0.002% max.
Assay Percent Range 99%
Specific Gravity 0.865
Linear Formula C6H4(CH3)2
Vapor Pressure Vapor Pressure: 8mbar at 20°C
Viscosity 0.6 mPa.s (20°C)
Water 0.05% max. (Coulometric)
Beilstein 05, 360
Merck Index 15, 10278
Boiling Point 136.0°C to 140.0°C
Solubility Information Solubility in water: <0.1g/L (20°C). Other solubilities: soluble in methanol, ethanol, 2-propanol, dmf,, methylene chloride, acetone and ether
Formula Weight 106.17
Percent Purity 99%
Grade ACS
Chemical Name or Material Xylenes
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.