missing translation for 'onlineSavingsMsg'
Learn More

Xylenes, puriss. p.a., ACS reagent, Honeywell

Product Code. 15674080
Change view
Click to view available options
Quantity:
1 L
2.5 L
25 L
144 x 1 L
Unit Size:
1 L
2.5 L
25 L
Pack of 144
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15674080 1 L 1 L
15684080 2.5 L 2.5 L
15694080 25 L 25 L
15664080 144 x 1 L Pack of 144
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
Product Code. 15674080 Supplier Honeywell Riedel-de Haën Supplier No. 338171L
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

Chemical Identifiers

CAS 1330-20-7
MDL Number MFCD00077264

Specifications

Density 0.86g/mL at 25°C
Quantity 1 L
Explosion Limit 0.07
Flash Point 25°C (77°F)
Linear Formula C6H4(CH3)2
Vapor Pressure 18 mmHg (37.7°C)
UN Number UN1307
Formula Weight 106.17g/mol
Grade ACS/puriss. p.a.
Chemical Name or Material Xylenes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.