missing translation for 'onlineSavingsMsg'
Learn More

Xylenes, PHOTREX™ Reagent, J.T. Baker™

Product Code. 15568624 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
500 mL
Packaging:
Narrow Mouth Amber Glass Bottle
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15568624 500 mL Narrow Mouth Amber Glass Bottle 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
Product Code. 15568624 Supplier Avantor J.T.Baker Supplier No. 951601

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

High purity solvents characterized for use in UV, visible and IR spectrophotometry

    For Spectrophotometry

Specifications

CAS 1330-20-7
Quantity 500 mL
Flash Point 29.00°C
Packaging Narrow Mouth Amber Glass Bottle
Specific Gravity 1L = 0.86kg
Molecular Formula C6H4(CH3)2
Formula Weight 106.17
Grade PHOTREX™ Reagent
Chemical Name or Material Xylenes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.