missing translation for 'onlineSavingsMsg'
Learn More

Xylenes, ACS, 98.5+% (Assay, isomers plus ethylbenzene)

Product Code. 11327188
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
4 x 1 L
Unit Size:
1 L
4 L
500mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11327188 1 L 1 L
11400490 500 mL 500mL
11307188 4 x 1 L 4 L
11317188 4 L 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
Product Code. 11327188 Supplier Thermo Scientific Alfa Aesar Supplier No. 016371.K2
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

CAS: 1330-20-7 | (CH3)2C6H4

Xylene is used as a solvent in pharmaceutical, printing, rubber, and leather industries. It is an important component of ink, rubber and adhesives as well as in thinning paints and varnishes. It is used in the laboratory to make baths with dry ice to cool reaction vessels, and as a solvent to remove synthetic immersion oil from the microscope objective in light microscopy. It finds application as a short-term hydrophobic clearing agent of tissue samples by histologists. It is a precursor for the preparation of xylene dichlorides, xylyl bromide, terephthalic acid and dimethyl terephthalate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1330-20-7
Molecular Formula (CH3)2C6H4
MDL Number MFCD00077264

Specifications

Melting Point <°C to 25°C
Color Colorless
Density 0.860
Quantity 1 L
Flash Point 29°C (84°F)
Physical Form Liquid
Refractive Index 1.497
Assay Percent Range >98.5%
Beilstein 1901563
Boiling Point 137°C to 144°C
Solubility Information Miscible with aromatic hydrocarbons. Immiscible with water.
Formula Weight 106.17
Grade ACS
Chemical Name or Material Xylenes, For ACS analysis
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Xylenes
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Aspiration toxicity Category 1
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity after repeated exposure Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H312-Harmful in contact with skin.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H335-May cause respiratory irritation.
  • H373-May cause damage to organs.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P331-Do NOT induce vomiting.
Supplemental information
  • MIXTURE LIST-Contain: Xylenes, Ethylbenzene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.