missing translation for 'onlineSavingsMsg'
Learn More

Xylene (mixture of isomers), BAKER ANALYZED™ ACS, J.T. Baker™

Product Code. 10221051 Shop All J.T.Baker Products
Change view
Click to view available options
:
5 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10221051 5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
Product Code. 10221051 Supplier JT Baker Supplier No. 8080.5000

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

Exceeds ACS Specifications

Specifications

Melting Point -34°C
CAS 1330-20-7
Color Max. 10 (APHA)
Density 0.86 - 0.88 g/cm³ (20°C)
Quantity 5 L
Flash Point 25°C
Residue after Evaporation max. 0.002%
Assay Percent Range min. 98.5%
Molecular Formula C8H10
MDL Number MFCD00077264
Water max. 0.03%
Boiling Point 140°C (1013 hPa)
InChI Key IVSZLXZYQVIEFR-UHFFFAOYSA-N
SMILES CC1=CC(C)=CC=C1
IUPAC Name 1,3-xylene
Molecular Weight (g/mol) 106.17
Show More Show Less
compliance-icons compliance-icons
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H312-Harmful in contact with skin.
  • H315-Causes skin irritation.
  • H332-Harmful if inhaled.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.