missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Xylene, for Electronic Use (MOS), Fisher Chemical™
Shop All Fisher Chemical Products
Click to view available options
Quantity:
2.5 L
5 L
Packaging:
Aluminum Can
Amber Glass Bottle
Unit Size:
2.5 L
5 L
Specifications
Specifications
| Melting Point | -34°C |
| Color | Colorless |
| Boiling Point | 136°C |
| Quantity | 2.5 L |
| Packaging | Amber Glass Bottle |
| Physical Form | Liquid |
| Vapor Pressure | 8mbar at 20°C |
| Viscosity | 0.6 mPaS at 20°C |
| Formula Weight | 106.17g/mol |
Safety and Handling
Product Identifier
- Xylene
- Danger
- Flammable liquid Category 3
- Aspiration toxicity Category 1
- Acute toxicity Category 4
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
- Specific target organ toxicity after repeated exposure Category 2
- LONG-TERM AQUATIC HAZARD Chronic 3
- H226-Flammable liquid and vapour.
- H304-May be fatal if swallowed and enters airways.
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H373-May cause damage to organs.
- H412-Harmful to aquatic life with long lasting effects.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P331-Do NOT induce vomiting.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P264-Wash face, hands and anyexposed skin thoroughly after handling.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction