missing translation for 'onlineSavingsMsg'
Learn More

Xylene Cyanol, pure

Product Code. 10463931 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
50 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
50g
5g
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10463931 5 g Glass bottle 5g
10118040 25 g Glass bottle 25g
10732022 50 g Glass bottle 50g
10178130 100 g Glass bottle 100g
10414511 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 10463931 Supplier Thermo Scientific Acros Supplier No. 422690050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 4463-44-9 | C25H28N2NaO7S2 | 555.616 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4463-44-9
Molecular Formula C25H28N2NaO7S2
Molecular Weight (g/mol) 555.616
MDL Number MFCD00019481
InChI Key JKPDSWLBCLGXEQ-UHFFFAOYSA-N
Synonym C.I. 43535, Cyanol FF
PubChem CID 131854036
IUPAC Name 4-[[4-(ethylamino)-3-methylphenyl]-(4-ethylimino-3-methylcyclohexa-2,5-dien-1-ylidene)methyl]-6-hydroxybenzene-1,3-disulfonic acid;sodium
SMILES CCNC1=C(C=C(C=C1)C(=C2C=CC(=NCC)C(=C2)C)C3=CC(=C(C=C3S(=O)(=O)O)S(=O)(=O)O)O)C.[Na]

Specifications

Dye Content 80% min.
Infrared Spectrum Authentic
Formula Weight 554.61
Grade Extra Pure
Packaging Glass bottle
Color Blue to Red
Quantity 5 g
Physical Form Crystalline Powder
Chemical Name or Material Xylene cyanol, 85%
compliance-icons
Product Identifier
  • Xylene cyanol
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.