missing translation for 'onlineSavingsMsg'
Learn More

Xylene, Certified AR for Analysis, Fisher Chemical™

Product Code. 10477270 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
5 L
25 L
Packaging:
Amber Glass Bottle
Metal Can
Plastic Canister
Plastic-coated Glass Bottle
Unit Size:
1 L
2.5 L
25 L
5 L
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10477270 25 L Metal Can 25 L
10467270 1 L Amber Glass Bottle 1 L
10385910 2.5 L Amber Glass Bottle 2.5 L
10418473 2.5 L Plastic-coated Glass Bottle 2.5 L
10112310 5 L Plastic Canister 5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
Product Code. 10477270 missing translation for 'mfr' Fisher Chemical missing translation for 'supplierNo' X/0250/25
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

C8H10, CAS Number-1330-20-7, 2.5L, 136 deg.C, Colorless, 106.17g/mol, -34 deg.C, 77264, Amber glass bottle, Liquid, 8mbar at 20 deg.C, 0.6 mPaS at 20 deg.C | CAS: 1330-20-7 | C8H10

C8H10, CAS Number-1330-20-7, 2.5L, 136 deg.C, Colorless, 106.17g/mol, -34 deg.C, 77264, Amber glass bottle, Liquid, 8mbar at 20 deg.C, 0.6 mPaS at 20 deg.C

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1330-20-7
Molecular Formula C8H10
MDL Number 77264

Specifications

Melting Point -34°C
Color Colorless
Boiling Point 136°C
Quantity 25 L
Packaging Metal Can
Physical Form Liquid
Vapor Pressure 8mbar at 20°C
Viscosity 0.6 mPaS at 20°C
Formula Weight 106.17g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.