missing translation for 'onlineSavingsMsg'
Learn More

Xylene, Certified AR for Analysis, Fisher Chemical™

Product Code. 10467270 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
5 L
25 L
Packaging:
Amber Glass Bottle
Metal Can
Plastic Canister
Plastic-coated Glass Bottle
Unit Size:
1 L
2.5 L
25 L
5 L
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10467270 1 L Amber Glass Bottle 1 L
10385910 2.5 L Amber Glass Bottle 2.5 L
10418473 2.5 L Plastic-coated Glass Bottle 2.5 L
10112310 5 L Plastic Canister 5 L
10477270 25 L Metal Can 25 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
Product Code. 10467270 Supplier Fisher Chemical Supplier No. X/0250/15
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

C8H10, CAS Number-1330-20-7, 2.5L, 136 deg.C, Colorless, 106.17g/mol, -34 deg.C, 77264, Amber glass bottle, Liquid, 8mbar at 20 deg.C, 0.6 mPaS at 20 deg.C | CAS: 1330-20-7 | C8H10

C8H10, CAS Number-1330-20-7, 2.5L, 136 deg.C, Colorless, 106.17g/mol, -34 deg.C, 77264, Amber glass bottle, Liquid, 8mbar at 20 deg.C, 0.6 mPaS at 20 deg.C

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1330-20-7
Molecular Formula C8H10
MDL Number 77264

Specifications

Melting Point -34°C
Color Colorless
Quantity 1 L
Packaging Amber Glass Bottle
Physical Form Liquid
Vapor Pressure 8mbar at 20°C
Viscosity 0.6 mPaS at 20°C
Boiling Point 136°C
Formula Weight 106.17g/mol
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Xylene
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 3
  • Aspiration toxicity Category 1
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H312-Harmful in contact with skin.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H335-May cause respiratory irritation.
  • H373-May cause damage to organs.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.