missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
A grade of chemicals for laboratory use, for organic synthesis, and for routine research usually controlled by physical criteria and often by assay (by GC).
Specifications
Specifications
| CAS | 1330-20-7 |
| Color | Colorless |
| Appearance | Passes test |
| Density | 0.86g/mL |
| Quantity | 25 L |
| Flash Point | 24°C |
| Packaging | Steel |
| Physical Form | Liquid |
| Molecular Formula | C8H10 |
| Linear Formula | (CH3)2C6H4 |
| Show More |
Safety and Handling
Signal Word
- Warning
- Acute toxicity Category 4
- Flammable liquid Category 3
- Skin corrosion/irritation Category 2
- H226-Flammable liquid and vapour.
- H312-Harmful in contact with skin.
- H315-Causes skin irritation.
- H332-Harmful if inhaled.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?