missing translation for 'onlineSavingsMsg'
Learn More

Xylazole hydrochloride, Dr. Ehrenstorfer

Product Code. 30312901
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30312901 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30312901 Supplier Dr. Ehrenstorfer Supplier No. DREC17943600

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Xylazole hydrochloride
CAS 123941-49-1
Analyte or Component Names Xylazole hydrochloride
Shipping Condition Room Temperature
Molecular Formula C11 H12 N2 S . Cl H
Certifications/Compliance ISO/IEC 17025
Synonym 2-Thiazolamine, N-(2,6-dimethylphenyl)-, hydrochloride (1:1), 2-Thiazolamine, N-(2,6-dimethylphenyl)-, monohydrochloride (9CI), Xylazole
InChI Formula InChI=1S/C11H12N2S.ClH/c1-8-4-3-5-9(2)10(8)13-11-12-6-7-14-11;/h3-7H,1-2H3,(H,12,13);1H
Solution Type Neat
SMILES Cl.Cc1cccc(C)c1Nc2nccs2
IUPAC Name N-(2,6-dimethylphenyl)-1,3-thiazol-2-amine;hydrochloride
Industry Type Environmental Testing
Molecular Weight (g/mol) 240.75
Quantity 100 mg
Formula Weight 240.0488 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.