missing translation for 'onlineSavingsMsg'
Learn More

Xylazine hydrochloride, MP Biomedicals™

Product Code. 11459791 Shop All MP Biomedicals Products
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g

Product Code. 11459791

Brand: MP Biomedicals 0215830725

Please to purchase this item. Need a web account? Register with us today!

Xylazine hydrochloride, White Powder

  • It has been used together to produce rapid and reversible anesthesia in experimental animals.
  • Xylazine elicited a diuretic response in ketamine-anesthesized animals, probably through activation of complex peripheral and CNS α2-adrenergic receptor systems.
  • It is an alpha agonist with sedative, analgesic and muscle relaxant properties.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23076-35-9
Molecular Weight (g/mol) 256.79
MDL Number MFCD00058196
InChI Key QYEFBJRXKKSABU-UHFFFAOYSA-N
Synonym 2-[2,6-Dimethylphenylamino]-4H-5,6-dihydroxthiazine, 5,6-Dihydro-2-(2,6-xylidino)-4H-1,3-thiazine hydrochloride
IUPAC Name hydrogen N-(2,6-dimethylphenyl)-5,6-dihydro-4H-1,3-thiazin-2-amine chloride
SMILES [H+].[Cl-].CC1=CC=CC(C)=C1NC1=NCCCS1

Specifications

Melting Point 161°C to 171°C
Color White
Loss on Drying 0.39
Molecular Formula C12H17ClN2S
Quantity 25 g
UN Number UN2811
Beilstein 6448471
Solubility Information Soluble in water (50mg/mL).
Physical Form Powder
Assay 1.0018
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.