missing translation for 'onlineSavingsMsg'
Learn More

Xylazine hydrochloride, MP Biomedicals™

Product Code. 11439791 Shop All MP Biomedicals Products
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g

Product Code. 11439791

Brand: MP Biomedicals 0215830701

Please to purchase this item. Need a web account? Register with us today!

Xylazine hydrochloride, White Powder

  • It has been used together to produce rapid and reversible anesthesia in experimental animals.
  • Xylazine elicited a diuretic response in ketamine-anesthesized animals, probably through activation of complex peripheral and CNS α2-adrenergic receptor systems.
  • It is an alpha agonist with sedative, analgesic and muscle relaxant properties.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

MDL Number MFCD00058196
Synonym 2-[2,6-Dimethylphenylamino]-4H-5,6-dihydroxthiazine, 5,6-Dihydro-2-(2,6-xylidino)-4H-1,3-thiazine hydrochloride

Specifications

Melting Point 161°C to 171°C
Color White
Loss on Drying 0.0039
Molecular Formula C12H16N2S . HCl
Quantity 1 g
UN Number UN2811
Beilstein 6448471
Solubility Information Soluble in water (50mg/mL).
Physical Form Powder
Assay 1.0018
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.