missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Xylazine Hydrochloride, LoGiCal
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Specifications
Specifications
| Chemical Name or Material | Xylazine hydrochloride |
| CAS | 23076-35-9 |
| Analyte or Component Names | Xylazine hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C12 H16 N2 S . Cl H |
| Certifications/Compliance | ISO 9001 |
| Synonym | Xylazine Hydrochloride, N-(2,6-Dimethylphenyl)-5,6-dihydro-4H-1,3-thiazin-2-amine monohydrochloride, 5,6-Dihydro-2-(2,6-xylidino)-4H-1,3-thiazine hydrochloride, BAY 1470 hydrochloride, BAY-Va 1470, Celactal, Narcosyl, Rompun, Xylapan, Xylasol, Xylazine chloride, Xylazine hydrochloride |
| InChI Formula | InChI=1S/C12H16N2S.ClH/c1-9-5-3-6-10(2)11(9)14-12-13-7-4-8-15-12;/h3,5-6H,4,7-8H2,1-2H3,(H,13,14);1H |
| Solution Type | Neat |
| SMILES | Cl.Cc1cccc(C)c1NC2=NCCCS2 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction