missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Xylazine-d6 Hydrochloride, TRC
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Specifications
Specifications
| Chemical Name or Material | Xylazine-d6 Hydrochloride |
| CAS | 1795020-86-8 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C12 D6 H10 N2 S . H Cl |
| Quantity | 10 mg |
| Synonym | N-[2,6-(Dimethyl-d6)phenyl]-5,6-dihydro-4H-1,3-thiazin-2-amine Hydrochloride, 5,6-Dihydro-2-(2,6-xylidino)-4H-1,3-thiazine-d6 Hydrochloride, 2-[2,6-(Dimethyl-d6)phenylamino]-5,6-dihydro-4H-1,3-thiazine Hydrochloride, BAY-Va 1470-d6, Celactal-d6, Narcosyl-d6, Rompun-d6, Xylapan-d6, Xylasol-d6 |
| InChI Formula | InChI=1S/C12H16N2S.ClH/c1-9-5-3-6-10(2)11(9)14-12-13-7-4-8-15-12;/h3,5-6H,4,7-8H2,1-2H3,(H,13,14);1H/i1D3,2D3; |
| SMILES | Cl.[2H]C([2H])([2H])c1cccc(c1NC2=NCCCS2)C([2H])([2H])[2H] |
| IUPAC Name | N-[2,6-bis(trideuteriomethyl)phenyl]-5,6-dihydro-4H-1,3-thiazin-2-amine;hydrochloride |
| Molecular Weight (g/mol) | 262.832 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction